Merck
  • Adductomic signatures of benzene exposure provide insights into cancer induction.
Adductomic signatures of benzene exposure provide insights into cancer induction.
Carcinogenesis (2018-03-15)
Hasmik Grigoryan, William M B Edmands, Qing Lan, Henrik Carlsson, Roel Vermeulen, Luoping Zhang, Song-Nian Yin, Gui-Lan Li, Martyn T Smith, Nathaniel Rothman, Stephen M Rappaport
ABSTRACT
Although benzene has long been recognized as a cause of human leukemia, the mechanism by which this simple molecule causes cancer has been problematic. A complicating factor is benzene metabolism, which produces many reactive intermediates, some specific to benzene and others derived from redox processes. Using archived serum from 20 nonsmoking Chinese workers, 10 with and 10 without occupational exposure to benzene (exposed: 3.2-88.9 ppm, controls: 0.002-0.020 ppm), we employed an adductomic pipeline to characterize protein modifications at Cys34 of human serum albumin, a nucleophilic hotspot in extracellular fluids. Of the 47 measured human serum albumin modifications, 39 were present at higher concentrations in benzene-exposed workers than in controls and many of the exposed-control differences were statistically significant. Correlation analysis identified three prominent clusters of adducts, namely putative modifications by benzene oxide and a benzene diolepoxide that grouped with other measures of benzene exposure, adducts of reactive oxygen and carbonyl species, and Cys34 disulfides of small thiols that are formed following oxidation of Cys34. Benzene diolepoxides are potent mutagens and carcinogens that have received little attention as potential causes of human leukemia. Reactive oxygen and carbonyl species-generated by redox processes involving polyphenolic benzene metabolites and by Cyp2E1 regulation following benzene exposure-can modify DNA and proteins in ways that contribute to cancer. The fact that these diverse human serum albumin modifications differed between benzene-exposed and control workers suggests that benzene can increase leukemia risks via multiple pathways involving a constellation of reactive molecules.
Sigma-Aldrich
3-Buten-2-one, contains 0.3-1.0% hydroquinone as stabilizer, technical grade, 90%
Sigma-Aldrich
Propionyl bromide, 97%
Sigma-Aldrich
Tiglic aldehyde, ≥96%