
Zirconium(IV) butoxide solution

Synonym(s):
Tetrabutyl zirconate solution
Linear Formula:
Zr(OC4H9)4
CAS Number:
Molecular Weight:
383.68
Products
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing