![Tris[N,N-bis(trimethylsilyl)amide]yttrium](/deepweb/assets/sigmaaldrich/product/structures/867/983/5b7cb7cd-8879-49e4-a9d7-29c52aaa82a0/640/5b7cb7cd-8879-49e4-a9d7-29c52aaa82a0.png)
Tris[N,N-bis(trimethylsilyl)amide]yttrium
![Tris[N,N-bis(trimethylsilyl)amide]yttrium](/deepweb/assets/sigmaaldrich/product/structures/867/983/5b7cb7cd-8879-49e4-a9d7-29c52aaa82a0/640/5b7cb7cd-8879-49e4-a9d7-29c52aaa82a0.png)
Linear Formula:
[[(CH3)3Si]2N]3Y
CAS Number:
Molecular Weight:
570.06
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing