Skip to Content
Merck
trans-2-Hexenoic acid 99%

trans-2-Hexenoic acid

trans-2-Hexenoic acid 99%
Linear Formula:
CH3CH2CH2CH=CHCO2H
CAS Number:
Molecular Weight:
114.14
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing