Skip to Content
Merck
Tolterodine Tartrate Pharmaceutical Secondary Standard: Certified Reference Material

Tolterodine tartrate

Tolterodine Tartrate Pharmaceutical Secondary Standard: Certified Reference Material
Synonym(s):
(R)-Tolterodine L-tartrate
Empirical Formula (Hill Notation):
C22H31NO · C4H6O6
CAS Number:
Molecular Weight:
475.57

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade