Skip to Content
Merck
Tinidazole Related Compound B certified reference material, pharmaceutical secondary standard

Tinidazole Related Compound B

Tinidazole Related Compound B certified reference material, pharmaceutical secondary standard
Synonym(s):
1-[2-(Ethylsulfonyl)ethyl]-2-methyl-4-nitro-1H-imidazole
Empirical Formula (Hill Notation):
C8H13N3O4S
CAS Number:
Molecular Weight:
247.27

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade