Skip to Content
CY
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Thymidine 5′-monophosphate disodium salt hydrate
Synonym(s):
5′-Thymidylic acid disodium salt, TMP, dTMP
Empirical Formula (Hill Notation):
C
10
H
13
N
2
Na
2
O
8
P · xH
2
O
CAS Number:
33430-62-5
Molecular Weight:
366.17 (anhydrous basis)
EC Number:
251-518-0
Products
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
T7004
Thymidine 5′-monophosphate disodium salt hydrate,
≥99%
Expand
View Pricing