
Tetrabenzyl pyrophosphate

Synonym(s):
Pyrophosphoric acid tetrabenzyl ester, Tetrabenzyl diphosphate
Linear Formula:
(C6H5CH2O)2P(O)OP(O)(OCH2C6H5)2
CAS Number:
Molecular Weight:
538.47
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing