Skip to Content
Merck
tBuXPhos 95%

tBuXPhos

tBuXPhos 95%
Synonym(s):
2-Di-tert-butylphosphino-2′,4′,6′-triisopropylbiphenyl, t-Bu XPhos, tert-Butyl XPhos
Empirical Formula (Hill Notation):
C29H45P
CAS Number:
Molecular Weight:
424.64

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing