Skip to Content
Merck
(S)-(−)-Perillic acid 95%

(S)-(−)-Perillic acid

(S)-(−)-Perillic acid 95%
Synonym(s):
4-Isopropenylcyclohexene-1-carboxylic acid
Empirical Formula (Hill Notation):
C10H14O2
CAS Number:
Molecular Weight:
166.22

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing