Skip to Content
Merck
Sodium L-lactate Pharmaceutical Secondary Standard; Certified Reference Material

Sodium L-lactate

Sodium L-lactate Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
(S)-2-Hydroxypropionic acid sodium salt, L-Lactic acid sodium salt, Sarcolactic acid sodium salt
Empirical Formula (Hill Notation):
C3H5NaO3
CAS Number:
Molecular Weight:
112.06
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3