
(S)-(−)-2-Methyl-CBS-oxazaborolidine solution

Synonym(s):
α,α-Diphenyl-L-prolinol methylboronic acid cycl-amide ester, (S)-1-Methyl-3,3-diphenyl-tetrahydro-pyrrolo[1,2c][1,3,2]oxazaborole, (S)-3,3-Diphenyl-1-methyltetrahydro-3H-pyrrolo[1,2-c][1,3,2]oxazaborole, (S)-Tetrahydro-1-methyl-3,3-diphenyl-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole
Empirical Formula (Hill Notation):
C18H20BNO
CAS Number:
Molecular Weight:
277.17
Products
Brand
Boiling Point (°C)
Formula Weight
Functional Group
Markush Class
Markush Group
Optical Isomer
Showing 1-2 of 2
Compare
Product Number
Description
Pricing