
Resomer® RG 503 H, Poly(D,L-lactide-co-glycolide)

Synonym(s):
Poly(D,L-lactide-co-glycolide), PLGA
Linear Formula:
[C3H4O2]x[C2H2O2]y
Products
Brand
Physical Form
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing