Skip to Content
Merck
R-(−)-Apomorphine hydrochloride hemihydrate calcined, ≥98% (with NaOH, titration)

R-(−)-Apomorphine hydrochloride hemihydrate

R-(−)-Apomorphine hydrochloride hemihydrate calcined, ≥98% (with NaOH, titration)
Linear Formula:
C17H17NO2 · HCl · 1/2H2O
CAS Number:
Molecular Weight:
312.79
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1