Skip to Content
Merck
Pyrantel pamoate VETRANAL®, analytical standard

Pyrantel pamoate

Pyrantel pamoate VETRANAL®, analytical standard
Synonym(s):
1-Methyl-2-(2-[2-thienyl]ethenyl)-1,4,5,6-tetrahydropyrimidine compound with 4,4′-methylenebis(3-hydroxy-2-naphthoic acid), Pyrantel embonate, Pyrantel pamoate
Linear Formula:
C11H14N2S · C23H16O6
CAS Number:
Molecular Weight:
594.68
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3