
Propentofylline

Synonym(s):
3,7-Dihydro-3-methyl-1-(5-oxohexyl)-7-propyl-1H-purine-2,6-dione, 3-Methyl-1-(5-oxohexyl)-7-propylxanthine, HWA 285
Empirical Formula (Hill Notation):
C15H22N4O3
CAS Number:
Molecular Weight:
306.36
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing