Skip to Content
Merck
Prednisolone acetate British Pharmacopoeia (BP) Reference Standard

Prednisolone acetate

Prednisolone acetate British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Prednisolone 21-acetate, 1,4-Pregnadiene-11β,17α,21-triol-3,20-dione 21-acetate, 11β,17α,21-Trihydroxy-1,4-pregnadiene-3,20-dione 21-acetate, 21-Acetoxy-1,4-pregnadiene-11β,17α-diol-3,20-dione
Empirical Formula (Hill Notation):
C23H30O6
CAS Number:
Molecular Weight:
402.48
EC Number:

Products

Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique