
Potassium bis(trimethylsilyl)amide solution

Synonym(s):
Hexamethyldisilazane potassium salt solution, Potassium hexamethyldisilazide solution
Linear Formula:
[(CH3)3Si]2NK
CAS Number:
Molecular Weight:
199.48
Products
Product Category
Brand
Sustainability
Color
Feature
Formula Weight
Physical Form
Quality Segment
Showing 1-3 of 3