
Potassium bicarbonate

Synonym(s):
Potassium hydrogen carbonate
Linear Formula:
KHCO3
CAS Number:
Molecular Weight:
100.12
Products
Product Category
Brand
Formula Weight
Manufacturer
pH Value
Physical Form
Product Line
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing