
Polyethylenimine hydrochloride

Synonym(s):
Linear PEI HCl salt, Poly(ethylene imine hydrochloride), Poly(ethylene imine) HCl salt
Linear Formula:
CH3(NHC2H4)nOH·xHCl
Products
Brand
Melting Point (°C)
Physical Form
Polymer Composition
Polymer Type
Showing 1-3 of 3