Skip to Content
Merck
Pentaethylene glycol di(p-toluenesulfonate) 95%

Pentaethylene glycol di(p-toluenesulfonate)

Pentaethylene glycol di(p-toluenesulfonate) 95%
Synonym(s):
3,6,9,12-Tetraoxatetradecane-1,14-diyl ditosylate, Pentaethylene glycol ditosylate
Linear Formula:
CH3C6H4SO3(CH2CH2O)5SO2C6H4CH3
CAS Number:
Molecular Weight:
546.65

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing