
Oxaliplatin

Synonym(s):
[SP-4-2-(1R-trans)]-(1,2-Cyclohexanediamine-N,N′)[ethanedioata(2--)-O,O’]platinum, Oxaliplatin
Empirical Formula (Hill Notation):
C8H14N2O4Pt
CAS Number:
Molecular Weight:
397.29
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4