Skip to Content
Merck
o-Tolidine analytical standard

o-Tolidine

o-Tolidine analytical standard
Synonym(s):
3,3′-Dimethylbenzidine, 4,4′-Bianisidine
Linear Formula:
[-C6H3(CH3)-4-NH2]2
CAS Number:
Molecular Weight:
212.29
EC Number:

Products

Product Category
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing