Skip to Content
Merck
Oleyl alcohol analytical standard

Oleyl alcohol

Oleyl alcohol analytical standard
Synonym(s):
Z-9-Dodecen-1-ol, cis-9-Octadecen-1-ol, Oleyl alcohol
Linear Formula:
CH3(CH2)7CH=CH(CH2)7CH2OH
CAS Number:
Molecular Weight:
268.48
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade