
N-Succinyl-Ala-Ala-Pro-Phe p-nitroanilide

Synonym(s):
N-Succinyl-L-alanyl-L-alanyl-L-prolyl-L-phenylalanine 4-nitroanilide, N-Succinyl-Ala-Ala-Pro-Phe-pNA
Empirical Formula (Hill Notation):
C30H36N6O9
CAS Number:
Molecular Weight:
624.64
Products
Product Category
Brand
Formula Weight
Gene Alias
Showing 1-1 of 1
Compare
Product Number
Description
Pricing