Skip to Content
Merck
N,O-Di-Boc-hydroxylamine 97%

N,O-Di-Boc-hydroxylamine

N,O-Di-Boc-hydroxylamine 97%
Synonym(s):
N,O-Bis(tert-butoxycarbonyl)hydroxylamine, tert-Butyl N-(tert-butoxycarbonyloxy)carbamate
Linear Formula:
(CH3)3COCONHOCOOC(CH3)3
CAS Number:
Molecular Weight:
233.26
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing