
N-Octanoyl-L-homoserine lactone

Synonym(s):
N-[(3S)-Tetrahydro-2-oxo-3-furanyl]octanamide, C8-HSL
Empirical Formula (Hill Notation):
C12H21NO3
CAS Number:
Molecular Weight:
227.30
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Quality Segment
Application
Optical Isomer
Amino Acid
Showing 1-2 of 2
Compare
Product Number
Description
Pricing