
N,N′-Di-Boc-1H-pyrazole-1-carboxamidine

Empirical Formula (Hill Notation):
C14H22N4O4
CAS Number:
Molecular Weight:
310.35
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing