
N-(Isobutoxymethyl)acrylamide

Linear Formula:
CH2=CHCONHCH2OCH2CH(CH3)2
CAS Number:
Molecular Weight:
157.21
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Monomer Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing