Skip to Content
Merck
Nickel carbonate, basic hydrate 99.9% trace metals basis

Nickel carbonate, basic hydrate

Nickel carbonate, basic hydrate 99.9% trace metals basis
Linear Formula:
NiCO3 · 2Ni(OH)2 · xH2O
CAS Number:
Molecular Weight:
304.12 (anhydrous basis)

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing