
Methyl tetracosanoate

Synonym(s):
Lignoceric acid methyl ester, Methyl tetracosanoate
Linear Formula:
CH3(CH2)22COOCH3
CAS Number:
Molecular Weight:
382.66
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Functional Group
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3