Skip to Content
Merck
Methyl-d3-magnesium iodide solution 1.0 M in diethyl ether, 99 atom % D

Methyl-d3-magnesium iodide solution

Methyl-d3-magnesium iodide solution 1.0 M in diethyl ether, 99 atom % D
Synonym(s):
Trideutero-methylmagnesium iodide
Linear Formula:
CD3MgI
CAS Number:
Molecular Weight:
169.26

Products

Product Category
Brand
Formula Weight
Physical Form
Isotope
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing