Skip to Content
Merck
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, inner salt 97%

(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, inner salt

(Methoxycarbonylsulfamoyl)triethylammonium hydroxide, inner salt 97%
Synonym(s):
Burgess reagent, Methyl N-(triethylammoniosulfonyl)carbamate
Linear Formula:
CH3O2CNSO2N(C2H5)3
CAS Number:
Molecular Weight:
238.30

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing