Methoxyallene
Synonym(s):
1-Methoxy-1,2-propadiene, Methyl propadienyl ether, NSC 363923
Empirical Formula (Hill Notation):
C4H6O
CAS Number:
Molecular Weight:
70.09
Products
Product Category
Brand
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing