
Metamizole Sodium

Synonym(s):
Metamizole sodium hydrate, (antipyrinylmethylamino)methanesulfonic acid sodium salt, 1-Phenyl-2,3-dimethyl-5-pyrazolone-4-methylaminomethanesulfonate sodium, 4-Methylamino-1,5-dimethyl-2-phenyl-3-pyrazolone sodium methanesulfonate, Analgin, Dipyrone, Metamizol, Methampyrone, Methylmelubrin, Noraminopyrine methanesulfonate sodium, Sodium methylaminoantipyrine methanesulfonate, Sulpyrine, [(2,3-Dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)methylamino]methanesulfonic acid sodium salt, Dipyrone monohydrate
Empirical Formula (Hill Notation):
C13H16N3NaO4S · H2O
CAS Number:
Molecular Weight:
351.35
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2