
Mesalazine impurity A

Synonym(s):
4-Aminophenol, 4-Hydroxyaniline
Linear Formula:
H2NC6H4OH
CAS Number:
Molecular Weight:
109.13
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing