
L-Cysteine hydrochloride

Synonym(s):
L-Cysteine hydrochloride monohydrate
Linear Formula:
HSCH2CH(NH2)COOH · HCl · H2O
CAS Number:
Molecular Weight:
175.63
Products
Formula Weight
Manufacturer
Physical Form
Format
Application
Markush Class
Markush Group
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing