
Kanamycin B sulfate salt

Synonym(s):
Bekanamycin sulfate salt
Empirical Formula (Hill Notation):
C18H37N5O10 · xH2SO4
Molecular Weight:
483.51 (free base basis)
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Format
Application
Mode of Action
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing