Isorhynchophylline
Synonym(s):
7-Isorhyncophylline
Empirical Formula (Hill Notation):
C22H28N2O4
CAS Number:
Molecular Weight:
384.47
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1