
Isoamyl butyrate

Synonym(s):
Isoamyl butyrate
Linear Formula:
CH3CH2CH2COOCH2CH2CH(CH3)2
CAS Number:
Molecular Weight:
158.24
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing