
Imidazole EMPROVE® EXPERT ACS

Synonym(s):
Imidazole
Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing