
Hydroxy naphthol blue disodium salt

Linear Formula:
C20H12N2O11S3Na2
CAS Number:
Molecular Weight:
598.49
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing