
Hexadecyltrimethylammonium p-toluenesulfonate

Synonym(s):
Cetyltrimethylammonium p-toluenesulfonate, Hexadecyltrimethylammonium-p-toluenesulfonate
Linear Formula:
CH3(CH2)15N(CH3C6H4SO3)(CH3)3
CAS Number:
Molecular Weight:
455.74
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing