
Glutathione

Synonym(s):
L-Glutathione reduced, γ-L-Glutamyl-L-cysteinyl-glycine, GSH
Linear Formula:
H2NCH(CO2H)CH2CH2CONHCH(CH2SH)CONHCH2CO2H
CAS Number:
Molecular Weight:
307.32
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique