
Fluorspar

Synonym(s):
Calcium fluoride, Fluorite, Fluorspar
Empirical Formula (Hill Notation):
CaF2
CAS Number:
Molecular Weight:
78.07
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing