Skip to Content
Merck
Fluoranthene-d10 98 atom % D

Fluoranthene-d10

Fluoranthene-d10 98 atom % D
Empirical Formula (Hill Notation):
C16D10
CAS Number:
Molecular Weight:
212.31

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing