Skip to Content
Merck
Ethyl 4-Nitrobenzoate Pharmaceutical Secondary Standard; Certified Reference Material

Ethyl 4-nitrobenzoate

Ethyl 4-Nitrobenzoate Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Ethyl 4-nitrobenzoate
Linear Formula:
O2NC6H4CO2C2H5
CAS Number:
Molecular Weight:
195.17

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade