
Erythromycin ethyl succinate

Synonym(s):
Erythromycin ethyl succinate
Empirical Formula (Hill Notation):
C43H75NO16
CAS Number:
Molecular Weight:
862.05
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing