
(±)-Methamphetamine solution

Empirical Formula (Hill Notation):
C10H15N
CAS Number:
Molecular Weight:
149.23
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing