Skip to Content
Merck
DL-Malic acid analytical standard

DL-Malic acid

DL-Malic acid analytical standard
Synonym(s):
(±)-2-Hydroxysuccinic acid, DL-Hydroxybutanedioic acid, DL-Malic acid
Linear Formula:
HO2CCH2CH(OH)CO2H
CAS Number:
Molecular Weight:
134.09
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Optical Isomer
Organoleptic Class
Reference Material Type
Special Grade
Technique